Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromothiophene

Catalog No. 106224100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-622-4100G 100 g
10-622-425G 25 g
10-622-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-622-4100G Supplier Sigma Aldrich Supplier No. 106224100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-bromothiophene can be used as a reactant to synthesize:
  • 3,3-bithiophene via borylation followed by suzuki coupling
  • 3-alkylthiophenes by nidppp++ catalyzed cross-coupling with grignard reagents
  • 3-lithiothiophene by treating with n-butyllithium in hexane
  • Derivatives of thienylenic α, -ddiformylαoligothiophenes
  • N-(2-(3-bromothiophen-2-yl)phenyl)methanesulfonamides by coupling at the 2-position with n-arylmethanesulfonamides, mediated by iodobenzene diacetate (178721)

Specifications

CAS 872-31-1
Density 1.74 g/mL (at 25°C (literature))
Boiling Point 150°C (lit.)
Molecular Formula C4H3BrS
Refractive Index n20/D 1.591 (literature)
Linear Formula C4H3BrS
MDL Number MFCD00005464
Quantity 100 g
Synonym 3-Thienyl bromide
Molecular Weight (g/mol) 163.04
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.