Learn More
Medchemexpress LLC 3-Bromothiophene | 872-31-1 | 99.6% | C4H3BrS | 1 G
SDP
Catalog No. 5000433783
Encompass_Preferred
3-Bromothiophene is a bromine atom-substituted 3-halogenated thiophene. Its polymers are conductive and hold potential optoelectronic application value. It serves as a key intermediate for synthesizing various important pharmaceuticals, agrochemicals, and cosmetics. Furthermore, it can be utilized in the synthesis of chalcones and their derivatives, some of which exhibit excellent anti-inflammatory, analgesic, and antibacterial activities.
  • Bromine atom-substituted 3-halogenated thiophene.
  • Polymers are conductive with potential optoelectronic application.
  • Key intermediate for synthesizing pharmaceuticals, agrochemicals, and cosmetics.
  • Used in the synthesis of chalcones and their derivatives.
  • Chalcone derivatives may exhibit anti-inflammatory, analgesic, and antibacterial activities.
Catalog No. 50-004-33783 Supplier Medchemexpress LLC Supplier No. HY592991G
3-Bromothiophene is a bromine atom-substituted 3-halogenated thiophene. Its polymers are conductive and hold potential optoelectronic application value. It serves as a key intermediate for synthesizing various important pharmaceuticals, agrochemicals, and cosmetics. Furthermore, it can be utilized in the synthesis of chalcones and their derivatives, some of which exhibit excellent anti-inflammatory, analgesic, and antibacterial activities.
  • Bromine atom-substituted 3-halogenated thiophene.
  • Polymers are conductive with potential optoelectronic application.
  • Key intermediate for synthesizing pharmaceuticals, agrochemicals, and cosmetics.
  • Used in the synthesis of chalcones and their derivatives.
  • Chalcone derivatives may exhibit anti-inflammatory, analgesic, and antibacterial activities.