Learn More
Medchemexpress LLC 3-Bromothiophene | 872-31-1 | 99.8% | C4H3BrS | 25 G
SDP
Catalog No. 5000358707
Encompass_Preferred
3-Bromothiophene is a bromine atom-substituted 3-halogenated thiophene. Its polymers are conductive and have potential optoelectronic application value. It also acts as a key intermediate for synthesizing various pharmaceuticals, agrochemicals, and cosmetics.
  • Bromine atom-substituted 3-halogenated thiophene
  • Polymers are conductive with optoelectronic application value
  • Key intermediate for synthesizing pharmaceuticals, agrochemicals, and cosmetics
  • Used in the synthesis of chalcones and their derivatives
  • Derivatives show anti-inflammatory, analgesic, and antibacterial activities
Catalog No. 50-003-58707 Supplier Medchemexpress LLC Supplier No. 25GHY59299
3-Bromothiophene is a bromine atom-substituted 3-halogenated thiophene. Its polymers are conductive and have potential optoelectronic application value. It also acts as a key intermediate for synthesizing various pharmaceuticals, agrochemicals, and cosmetics.
  • Bromine atom-substituted 3-halogenated thiophene
  • Polymers are conductive with optoelectronic application value
  • Key intermediate for synthesizing pharmaceuticals, agrochemicals, and cosmetics
  • Used in the synthesis of chalcones and their derivatives
  • Derivatives show anti-inflammatory, analgesic, and antibacterial activities