Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Buten-1-ol

Catalog No. 496839100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-683-9100G 100 g
49-683-91KG 1 kg
49-683-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 49-683-9100G Supplier Sigma Aldrich Supplier No. 496839100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-buten-1-ol is a homoallyl alcohol that can be prepared by the dehydration of 1,4-butanediol using cerium catalyst
  • The intramolecular hydrogen bonding of 3-buten-1-ol has been studied using ft-IR and 1h NMR spectroscopic data
  • Its microwave spectrum has been recorded and analyzed
  • The alkylation reaction of 3-buten-1-ol using titanium-organoaluminum system has been studied
  • Its gas-phase enthalpy of formation has been reported to be -147.3 +/- 1.8kj mol-1.employed in a study of the mn-catalyzed hydrohydrazination of olefins
  • Also used in a study of the conversion of propargylic acetates to ethers cataylzed by ferric chloride

Specifications

CAS 627-27-0
Density 0.838 g/mL (at 25°C (literature))
Boiling Point 112°C to 114°C (lit.)
Molecular Formula C4H8O
Refractive Index n20/D 1.421 (literature)
Linear Formula CH2=CHCH2CH2OH
MDL Number MFCD00002959
Quantity 100 g
Synonym Allylcarbinol
Molecular Weight (g/mol) 72.11
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.