Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Buten-1-ol, Spectrum™ Chemical
SDP

Catalog No. 18620240 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-240 100 mL
18-620-239 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-240 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1452100ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 627-27-0

Specifications

Boiling Point 114°C (237°F)
Quantity 100 mL
Molecular Weight (g/mol) 72.11
Percent Purity 96%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material 3-Buten-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.