Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Buten-1-yl Isothiocyanate 96.0+%, TCI America™
SDP

Catalog No. I04431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04431G Supplier TCI America Supplier No. I04431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3386-97-8
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00191660
InChI Key SKIHGKNFJKJXPX-UHFFFAOYSA-N
Synonym Isothiocyanic Acid 3-Buten-1-yl Ester
PubChem CID 76922
IUPAC Name 4-isothiocyanatobut-1-ene
SMILES C=CCCN=C=S

Specifications

Color Yellow
Boiling Point 77°C
Quantity 1 g
UN Number 1992
Formula Weight 113.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Buten-1-yl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.