Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Buten-2-one

Catalog No. M87509100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M87509100ML 100 mL
M87509500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M87509100ML Supplier Sigma Aldrich Supplier No. M87509100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-buten-2-one can undergo conjugate addition with methanol over the solid base catalysts to afford 4-methoxy-butan-2-one
  • It can also undergo coupling reaction with alkyltins and alkynes in the presence of a nickel complex to form stereo-defined conjugated enynes

Specifications

CAS 78-94-4
Density 0.864 g/mL (at 25°C (literature))
Boiling Point 81°C (lit.)
Molecular Formula C4H6O
Refractive Index n20/D 1.411 (literature)
Linear Formula CH2=CHCOCH3
MDL Number MFCD00008777
Quantity 100 mL
Synonym Methyl vinyl ketone; Vinyl methyl ketone
Molecular Weight (g/mol) 70.09
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.