Learn More
Medchemexpress LLC 3-buten-2-one, 4-(2-naphthalenyl)-, (3E)- | 51557-10-9 | 99.9% | 196.24 | 1 G
SDP
Catalog No. 5000403815
Encompass_Preferred
(E)-4-(Naphthalen-2-yl)but-3-en-2-one is a small active molecule that serves as a versatile building block. This compound demonstrates significant anti-leukemic activity, effectively inhibiting the proliferation of K562 human chronic myeloid leukemia cells.
  • Small active molecule
  • Can be used as a building block
  • Exhibits anti-leukemic activity
  • Inhibits proliferation of K562 cells with an IC50 of 7.6 μM
Catalog No. 50-004-03815 Supplier Medchemexpress LLC Supplier No. HY4028151G
(E)-4-(Naphthalen-2-yl)but-3-en-2-one is a small active molecule that serves as a versatile building block. This compound demonstrates significant anti-leukemic activity, effectively inhibiting the proliferation of K562 human chronic myeloid leukemia cells.
  • Small active molecule
  • Can be used as a building block
  • Exhibits anti-leukemic activity
  • Inhibits proliferation of K562 cells with an IC50 of 7.6 μM