Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Buten-2-Yl 1H-Imidazole-1-Carboxylate

Catalog No. 7528861G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-288-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 75-288-61G Supplier Sigma Aldrich Supplier No. 7528861G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Buten-2-Yl 1H-Imidazole-1-Carboxylate

Specifications

CAS 850568-55-7
Melting Point 254.1°C to 262.7°C
Molecular Formula C13H20BNO2 . HCl
Refractive Index n20/D 1.482
Linear Formula C13H20BNO2 . HCl
MDL Number MFCD02179455
Quantity 1 g
Synonym Heller-Sarpong Reagent; 1H-Imidazole-1-carboxylic acid 1-methyl-2-propen-1-yl ester; Sarpong 3-butenyl transesterification reagent; Sarpong reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.