Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-butenoic acid | 625-38-7 | 98.6% | 86.09 | C4H6O2 | 50 G
SDP

Catalog No. 5000209807
Encompass_Preferred

3-Butenoic acid is a small alkenoic acid used as a chemical intermediate in organic synthesis and pharmaceutical research. It is a colorless to light yellow liquid typically supplied at high purity for research applications.

  • Used as a chemical and drug intermediate
  • Liquid, colorless to light yellow
  • Molecular formula C4H6O2, molecular weight 86.09 g/mol
  • High purity for research (98.61%)
  • Available in multiple pack sizes including 50 g

Catalog No. 50-002-09807 Supplier Medchemexpress LLC Supplier No. HYW06869750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Butenoic acid is a small alkenoic acid used as a chemical intermediate in organic synthesis and pharmaceutical research. It is a colorless to light yellow liquid typically supplied at high purity for research applications.

  • Used as a chemical and drug intermediate
  • Liquid, colorless to light yellow
  • Molecular formula C4H6O2, molecular weight 86.09 g/mol
  • High purity for research (98.61%)
  • Available in multiple pack sizes including 50 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.