Learn More
Medchemexpress LLC 3-Butenoic acid | 625-38-7 | 98.6% | 86.09 g/mol | C4H6O2 | 1 ML
SDP
Catalog No. 5000210818
Encompass_Preferred
3-Butenoic acid (vinylacetic acid) is supplied as a ready-to-use 10 mM solution in DMSO, intended for research and synthetic applications as a small-molecule building block and analytical reagent. It is provided in a 1 mL vial for convenient use in assays or synthesis.
  • 10 mM solution in DMSO, 1 mL vial
  • High assay purity suitable for research applications
  • Used as a chemical intermediate and analytical reagent
  • Molecular formula C4H6O2 and molecular weight 86.09 g/mol
  • Provided as a solution to simplify handling and reconstitution
Catalog No. 50-002-10818 Supplier Medchemexpress LLC Supplier No. HYW06869710MM/1ML
3-Butenoic acid (vinylacetic acid) is supplied as a ready-to-use 10 mM solution in DMSO, intended for research and synthetic applications as a small-molecule building block and analytical reagent. It is provided in a 1 mL vial for convenient use in assays or synthesis.
  • 10 mM solution in DMSO, 1 mL vial
  • High assay purity suitable for research applications
  • Used as a chemical intermediate and analytical reagent
  • Molecular formula C4H6O2 and molecular weight 86.09 g/mol
  • Provided as a solution to simplify handling and reconstitution