Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. B173725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B173725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B173725ML Supplier TCI America Supplier No. B173725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1576-84-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00086596
InChI Key IEKXSSZASGLISC-UHFFFAOYSA-N
Synonym 4-Acetoxy-1-butene, Acetic Acid 3-Butenyl Ester
PubChem CID 74095
IUPAC Name but-3-enyl acetate
SMILES CC(=O)OCCC=C

Specifications

Color Colorless
Boiling Point 128°C
Quantity 25 mL
UN Number 3271
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butenyl Acetate
EINECSNumber (2)-0760
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.