Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butenylamine, 97%

Catalog No. AC455050010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455050010 1 g
AC455050050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455050010 Supplier Thermo Scientific Chemicals Supplier No. 455050010
  • $223.00
    $157.65 / Each of 1

    Save $65.35

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 2524-49-4 | C4H9N | 71.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-49-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD03425859
InChI Key ASVKKRLMJCWVQF-UHFFFAOYSA-N
Synonym 3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene
PubChem CID 443732
ChEBI CHEBI:31108
IUPAC Name but-3-en-1-amine
SMILES NCCC=C

Specifications

Boiling Point 83.0°C
Flash Point −5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula H2C=CH(CH2)2NH2
Quantity 1 g
Solubility Information Solubility in water: soluble.
Formula Weight 71.12
Percent Purity 97%
Chemical Name or Material 3-Butenylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (or ha

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.