Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog No. AAH58171AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58171AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58171AD Supplier Thermo Scientific Chemicals Supplier No. H58171AD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226570-67-8
Molecular Formula C4H7BrZn
Molecular Weight (g/mol) 200.384
MDL Number MFCD09801449
InChI Key BFFDFNINBXPQIU-UHFFFAOYSA-M
Synonym 3-butenylzinc bromide, bromo but-3-en-1-yl zinc, but-3-enylzinc bromide, 0.50 m in thf, 3-butenylzinc bromide 0.5m in tetrahydrofuran, 3-butenylzinc bromide solution, 0.6 m in thf, 3-butenylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 24722678
IUPAC Name bromozinc(1+);but-1-ene
SMILES [CH2-]CC=C.[Zn+]Br

Specifications

Linear Formula H2C=CH(CH2)2ZnBr
Quantity 50 mL
UN Number UN3399
Formula Weight 200.38
Chemical Name or Material 3-Butenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.