Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B46405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4640-5G 5 g
B46401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4640-5G Supplier TCI America Supplier No. B46405G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 352534-81-7
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.037
MDL Number MFCD03427053
InChI Key ZTRKYYYDXARQRD-UHFFFAOYSA-N
Synonym 3-Butoxybenzeneboronic Acid
PubChem CID 5084101
IUPAC Name (3-butoxyphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)OCCCC)(O)O

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Formula Weight 194.04
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.