Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Butyl-5-hydroxyphthalate | 195142-72-4 | 206.24 | 1 ML
SDP

Catalog No. 5000416317
Encompass_Preferred

3-Butyl-5-hydroxyphthalate is a natural product intended for research use only. With a purity of 99.82%, it appears as a white to off-white solid.

  • Purity: 99.82%
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C12H14O3
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-16317 Supplier Medchemexpress LLC Supplier No. HYN1176010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Butyl-5-hydroxyphthalate is a natural product intended for research use only. With a purity of 99.82%, it appears as a white to off-white solid.

  • Purity: 99.82%
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C12H14O3
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.