Learn More
TARGETMOL CHEMICALS INC 3-BUTYLIDENEPHTHALIDE 50MG
SDP
Catalog No. 502576834
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. 3-Butylidenephthalide (Butylidene phthalide) has antihyperglycemic effect is due to inhibition of (alpha)-glucosidase at the intestinal level inhibited the activity of yeast-(alpha)-glucosidase (IC(5) 2.35 mM) in a noncompetitive fashion with a K(i) of 4.86 mM. purity: 99%
Catalog No. 50-257-6834 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S207250MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. 3-Butylidenephthalide (Butylidene phthalide) has antihyperglycemic effect is due to inhibition of (alpha)-glucosidase at the intestinal level inhibited the activity of yeast-(alpha)-glucosidase (IC(5) 2.35 mM) in a noncompetitive fashion with a K(i) of 4.86 mM. purity: 99%