Learn More
Medchemexpress LLC HY-N0336 5mg Medchemexpress, 3-Butylidenephthalide CAS:551-08-6 Purity:>98%
SDP
Catalog No. 501970254
Encompass_Preferred
Medchemexpress, HY-N0336 5mg 3-Butylidenephthalide CAS:551-08-6 3-Butylidenephthalide (Butylidenephthalide) is a phthalic anhydride derivative identified in Ligusticum chuanxiong Hort, and has larvicidal activity (LC50 of 1.56 mg/g for Spodoptera litura larvae). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0254 Supplier Medchemexpress LLC Supplier No. HYN03365MG
Medchemexpress, HY-N0336 5mg 3-Butylidenephthalide CAS:551-08-6 3-Butylidenephthalide (Butylidenephthalide) is a phthalic anhydride derivative identified in Ligusticum chuanxiong Hort, and has larvicidal activity (LC50 of 1.56 mg/g for Spodoptera litura larvae). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.