Learn More
AdipoGen 3-Butyn-2-ol
SDP
Catalog No. 502561671
Encompass_Preferred
Chemical. CAS 2028-63-9. Formula C4H6O. MW 70.09. Synthetic. Organic building block. 3-Butyn-2-ol is an intermediate used to prepare aminoindanes with inhibitory activity toward acetylcholine esterase and monoamine oxidase useful as anti-Alzheimer’s agents.
Catalog No. 50-256-1671 Supplier AdipoGen Supplier No. CDXB0123G050
Chemical. CAS 2028-63-9. Formula C4H6O. MW 70.09. Synthetic. Organic building block. 3-Butyn-2-ol is an intermediate used to prepare aminoindanes with inhibitory activity toward acetylcholine esterase and monoamine oxidase useful as anti-Alzheimer’s agents.