Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butyn-2-ol (55% in Water, ca. 7.5mol/L), TCI America™
SDP

Catalog No. B075025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B075025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B075025ML Supplier TCI America Supplier No. B075025ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2028-63-9
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00004541
InChI Key GKPOMITUDGXOSB-UHFFFAOYNA-N
Synonym 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol
PubChem CID 16239
IUPAC Name but-3-yn-2-ol
SMILES CC(O)C#C

Specifications

Color Yellow
UN Number 2929
Quantity 25 mL
Formula Weight 70.09
Physical Form Liquid
Chemical Name or Material 3-Butyn-2-ol (55% in Water, ca. 7.5mol/L)
RTECSNumber ES0709800
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.