Learn More
Medchemexpress LLC 3-Butynoic acid | 2345-51-9 | 84.07 | 10 MM 1 ML
SDP
Catalog No. 5000448521
Encompass_Preferred
3-Butynoic acid is an inhibitor of acyl CoA dehydrogenase and a click chemistry reagent. It contains an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This chemical can reduce the formation of propionate and acrylic acid in a concentration-dependent manner. It is supplied as a 10 mM solution in DMSO and is intended for laboratory use.
  • Inhibits acyl CoA dehydrogenase.
  • Acts as a click chemistry reagent.
  • Reduces propionate and acrylic acid concentration-dependently.
  • Stable under recommended storage conditions.
  • Available as a human endogenous metabolite.
Catalog No. 50-004-48521 Supplier Medchemexpress LLC Supplier No. HYW00478410MM/1ML
3-Butynoic acid is an inhibitor of acyl CoA dehydrogenase and a click chemistry reagent. It contains an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This chemical can reduce the formation of propionate and acrylic acid in a concentration-dependent manner. It is supplied as a 10 mM solution in DMSO and is intended for laboratory use.
  • Inhibits acyl CoA dehydrogenase.
  • Acts as a click chemistry reagent.
  • Reduces propionate and acrylic acid concentration-dependently.
  • Stable under recommended storage conditions.
  • Available as a human endogenous metabolite.