Learn More
Medchemexpress LLC 3-Butynoic acid | 2345-51-9 | 84.07 | 250 MG
SDP
Catalog No. 5000449885
Encompass_Preferred
3-Butynoic acid is an inhibitor of acyl CoA dehydrogenase. It functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This compound can concentration-dependently reduce the formation of propionate and acrylic acid.
  • Inhibits acyl CoA dehydrogenase
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Reduces the formation of propionate and acrylic acid
  • Appears as a white to yellow solid
  • Identified as an endogenous metabolite
Catalog No. 50-004-49885 Supplier Medchemexpress LLC Supplier No. 250MGHYW004784
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3-Butynoic acid is an inhibitor of acyl CoA dehydrogenase. It functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. This compound can concentration-dependently reduce the formation of propionate and acrylic acid.
  • Inhibits acyl CoA dehydrogenase
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Reduces the formation of propionate and acrylic acid
  • Appears as a white to yellow solid
  • Identified as an endogenous metabolite