Learn More
Medchemexpress LLC 3-Butynoic acid | 2345-51-9 | MFCD02258494 | 84.07 | 500 MG
SDP
Catalog No. 5000446056
Encompass_Preferred
3-Butynoic acid is an inhibitor of acyl CoA dehydrogenase. It is also a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It can reduce the formation of propionate and acrylic acid concentration-dependently.
  • Inhibitor of acyl CoA dehydrogenase
  • Click chemistry reagent with alkyne group
  • Capable of CuAAc with azide-containing molecules
  • Reduces propionate and acrylic acid formation concentration-dependently
Catalog No. 50-004-46056 Supplier Medchemexpress LLC Supplier No. HYW004784500MG
3-Butynoic acid is an inhibitor of acyl CoA dehydrogenase. It is also a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It can reduce the formation of propionate and acrylic acid concentration-dependently.
  • Inhibitor of acyl CoA dehydrogenase
  • Click chemistry reagent with alkyne group
  • Capable of CuAAc with azide-containing molecules
  • Reduces propionate and acrylic acid formation concentration-dependently