Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Butynoic Acid 95.0+%, TCI America™
SDP

Catalog No. B4969200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4969200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4969200MG Supplier TCI America Supplier No. B4969200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2345-51-9
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.074
MDL Number MFCD02258494
InChI Key KKAHGSQLSTUDAV-UHFFFAOYSA-N
PubChem CID 137547
ChEBI CHEBI:1461
IUPAC Name but-3-ynoic acid
SMILES C#CCC(=O)O

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 200 mg
UN Number 3261
Formula Weight 84.07
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Butynoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.