Learn More
Medchemexpress LLC 3-C12-NBD-cholesterol | 1246303-05-8 | 99.9% | 747.06 | 1 MG
SDP
Catalog No. 5000352416
Encompass_Preferred
3-C12-NBD-cholesterol is a fluorescent cholesterol analog used for measuring membrane and intracellular trafficking dynamics. This product is a colorless to light yellow liquid with a purity of 99.9%.
  • Fluorescent cholesterol analog
  • Measures membrane and intracellular trafficking dynamics
  • High purity of 99.9%
  • Suitable for cell staining analysis
  • Used as an organelle dye
  • Used as a lipid droplet dye
  • Solution storage at -20°C for up to 2 years
Catalog No. 50-003-52416 Supplier Medchemexpress LLC Supplier No. HY1394071MG
3-C12-NBD-cholesterol is a fluorescent cholesterol analog used for measuring membrane and intracellular trafficking dynamics. This product is a colorless to light yellow liquid with a purity of 99.9%.
  • Fluorescent cholesterol analog
  • Measures membrane and intracellular trafficking dynamics
  • High purity of 99.9%
  • Suitable for cell staining analysis
  • Used as an organelle dye
  • Used as a lipid droplet dye
  • Solution storage at -20°C for up to 2 years