Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-campholenyl-2-butanol | 65113-99-7 | 98.0% | 210.36 | C14H26O | 5 MG
SDP

Catalog No. 5000222718
Encompass_Preferred

3-Campholenyl-2-butanol is a synthetic sandalwood-like odorant and a selective agonist of the olfactory receptor OR2AT4, used in biochemical and pharmacological research. It is provided at high purity and is suitable for receptor activation assays and related studies.

  • Purity 98.0%.
  • Molecular formula C14H26O; molecular weight 210.36 g/mol.
  • Available in small quantities including 5 mg and as 10 mM solutions in DMSO.
  • Soluble in DMSO for stock solutions and assay use.
  • Useful for olfactory receptor and hair growth-related functional studies.

Catalog No. 50-002-22718 Supplier Medchemexpress LLC Supplier No. HY1397835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Campholenyl-2-butanol is a synthetic sandalwood-like odorant and a selective agonist of the olfactory receptor OR2AT4, used in biochemical and pharmacological research. It is provided at high purity and is suitable for receptor activation assays and related studies.

  • Purity 98.0%.
  • Molecular formula C14H26O; molecular weight 210.36 g/mol.
  • Available in small quantities including 5 mg and as 10 mM solutions in DMSO.
  • Soluble in DMSO for stock solutions and assay use.
  • Useful for olfactory receptor and hair growth-related functional studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.