Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carbamoylbenzoic acid, Thermo Scientific™

Catalog No. AC469610010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469610010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469610010 Supplier Thermo Scientific Chemicals Supplier No. 469610010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material 3-Carbamoylbenzoic acid
Alpha Vector CARBAMOYLBENZOICACID
CAS 4481-28-1
Melting Point 280°C to 290°C
Color Beige to White
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C8 H7 N O3
Quantity 1 g
MDL Number 04118098
Synonym Isophthalamic acid
Molecular Weight (g/mol) 165.15
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.