Learn More
Medchemexpress LLC 3-Carboxy-1-(methyl-d3)pyridin-1-ium chloride | 6138-41-6 | 176.62 | 1 MG
SDP
Catalog No. 5000389945
Encompass_Preferred
Trigonelline-d3 (chloride) is the deuterium labeled Trigonelline chloride. Trigonelline chloride, an alkaloid with potential antidiabetic activity, is present in considerable amounts in coffee. Trigonelline chloride has anti-HSV-1, antibacterial, and antifungal activities.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89945 Supplier Medchemexpress LLC Supplier No. HYN0415S1MG
Trigonelline-d3 (chloride) is the deuterium labeled Trigonelline chloride. Trigonelline chloride, an alkaloid with potential antidiabetic activity, is present in considerable amounts in coffee. Trigonelline chloride has anti-HSV-1, antibacterial, and antifungal activities.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.