Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-carboxy-6-hydroxycoumarin | 56437-16-2 | MFCD16295011 | 99.9% | 206.15 g/mol | C10H6O5 | 1 ML
SDP

Catalog No. 5000299601
Encompass_Preferred

3-Carboxy-6-hydroxycoumarin is a coumarin-derived intermediate used in organic synthesis and chemical research. It is supplied in both solid form and as a 10 mM solution in DMSO for use in screening, synthesis, and analytical workflows.

  • High purity, typically around 99.9%.
  • Molecular formula C10H6O5 and molecular weight 206.15 g/mol.
  • Available as a white to yellow solid and as a 10 mM solution in DMSO.
  • Solid pack sizes commonly include 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg.
  • Storage recommendations: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-99601 Supplier Medchemexpress LLC Supplier No. HYW02302310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Carboxy-6-hydroxycoumarin is a coumarin-derived intermediate used in organic synthesis and chemical research. It is supplied in both solid form and as a 10 mM solution in DMSO for use in screening, synthesis, and analytical workflows.

  • High purity, typically around 99.9%.
  • Molecular formula C10H6O5 and molecular weight 206.15 g/mol.
  • Available as a white to yellow solid and as a 10 mM solution in DMSO.
  • Solid pack sizes commonly include 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg.
  • Storage recommendations: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent, store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.