Learn More
Apexbio Technology LLC 3-Carboxy-7-Hydroxycoumarin 779-27-1 1g
SDP
Catalog No. 5000093472
Encompass_Preferred
3-Carboxy-7-Hydroxycoumarin (CAS 779-27-1) is a coumarin-based fluorescent molecule It is characterized by blue fluorescence emission upon excitation with ultraviolet light Structurally it features a carboxyl group that enables conjugation via carbodiimide activation (e g EDAC-mediated formation of succinimidyl ester intermediates) allowing covalent attachment to amine-containing residues in proteins and nucleic acids Based on these properties 3-Carboxy-7-Hydroxycoumarin is commonly employed in biochemical research for fluorescent labeling including analysis of protein protein interactions nucleic acid visualization and biomolecule detection assays
Catalog No. 50-000-93472 Supplier Apexbio Technology LLC Supplier No. A47311000
3-Carboxy-7-Hydroxycoumarin (CAS 779-27-1) is a coumarin-based fluorescent molecule It is characterized by blue fluorescence emission upon excitation with ultraviolet light Structurally it features a carboxyl group that enables conjugation via carbodiimide activation (e g EDAC-mediated formation of succinimidyl ester intermediates) allowing covalent attachment to amine-containing residues in proteins and nucleic acids Based on these properties 3-Carboxy-7-Hydroxycoumarin is commonly employed in biochemical research for fluorescent labeling including analysis of protein protein interactions nucleic acid visualization and biomolecule detection assays