Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxy-proxyl, 98%, free radical, Thermo Scientific™

Catalog No. AC224890250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC224890250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC224890250 Supplier Thermo Scientific Chemicals Supplier No. 224890250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Polymerization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2154-68-9
Molecular Formula C9H16NO3
MDL Number MFCD00003167
Synonym pca radical, ccris 4554, 2,2,5,5-tetramethyl-3-carboxypyrrolidinooxy, 2,2,5,5-tetramethyl-3-carboxypyrrolidine-n-oxyl, 3-carboxy-2,2,5,5-tetramethylpyrrolidin-1-yloxy, 3-carboxy-2,2,5,5-tetramethylpyrrolidinyloxyl, 3-carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 3-carboxy-2,2,5,5-tetramethylpyrrolidine 1-oxyl, 3-carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl, 2,2,5,5-tetramethyl-1-pyrrolidinyloxy-3-carboxylic acid

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 mg
Formula Weight 186.23
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-Carboxy-proxyl, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.