Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxybenzaldehyde, 97%

Catalog No. AAB2127714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2127714 25 g
AAB2127703 1 g
AAB2127706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2127714 Supplier Thermo Scientific Chemicals Supplier No. B2127714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 619-21-6 | C8H6O3 | 150.133 g/mol

3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

Solubility
Soluble in methanol (100 mg/mL) gives a clear to slightly hazy, colorless to very faintly brown.

Notes
Air sensitive. Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-21-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00039575
InChI Key UHDNUPHSDMOGCR-UHFFFAOYSA-N
Synonym 3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
PubChem CID 12077
IUPAC Name 3-formylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C=O

Specifications

Melting Point 173°C to 175°C
Quantity 25 g
Beilstein 2206413
Solubility Information Soluble in methanol (100mg/mL) gives a clear to slightly hazy,colorless to very faintly brown.
Formula Weight 150.13
Percent Purity 97%
Chemical Name or Material 3-Carboxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-585-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.