Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic Acid 1,4:2,3-Dianhydride 97.0+%, TCI America™
SDP

Catalog No. C31101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31101G 1 g
C31105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31101G Supplier TCI America Supplier No. C31101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6053-46-9
Molecular Formula C10H8O6
Molecular Weight (g/mol) 224.168
MDL Number MFCD22373052
InChI Key QVEIRZNRYOJFCL-UHFFFAOYSA-N
PubChem CID 22239650
SMILES C1C2C3CC(=O)OC(=O)C3C1C(=O)OC2=O

Specifications

Melting Point 201°C
Color White
Quantity 1 g
Formula Weight 224.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Carboxymethyl)-1,2,4-cyclopentanetricarboxylic Acid 1,4:2,3-Dianhydride
EINECSNumber (5)-5988
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.