Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C20281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20281G 1 g
C2028-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20281G Supplier TCI America Supplier No. C20281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25487-66-5
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.94
MDL Number MFCD00036833
InChI Key DBVFWZMQJQMJCB-UHFFFAOYSA-N
PubChem CID 2733957
IUPAC Name 3-(dihydroxyboranyl)benzoic acid
SMILES OB(O)C1=CC=CC(=C1)C(O)=O

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 1 g
Synonym 3-carboxyphenylboronic acid, 3-dihydroxyboranyl benzoic acid, 3-carboxybenzeneboronic acid, m-carboxyphenylboronic acid, 3-dihydroxyborane benzoic acid, benzoic acid, 3-borono, 3-dihydroxyboryl benzoic acid, 3-phenyl ester boronic acid, 3-carboxyphenyl boronic acid, 3-carboxy-phenyl-boronic acid
Formula Weight 165.94
Physical Form Crystalline Powder
Chemical Name or Material 3-Carboxyphenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber CY8750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.