Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Carboxypropyl)triphenylphosphonium bromide, 97%

Catalog No. AC338930250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338930250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338930250 Supplier Thermo Scientific Chemicals Supplier No. 338930250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 17857-14-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 247.0°C to 249.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C22H22BrO2P
Quantity 25 g
MDL Number MFCD00274196
Synonym 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m
InChI Key NKVJKVMGJABKHV-UHFFFAOYSA-N
SMILES [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 3-carboxypropyl(triphenyl)phosphanium;bromide
Molecular Weight (g/mol) 429.29
PubChem CID 10717451
Formula Weight 429.29
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 3-(Carboxypropyl)triphenylphosphonium bromide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.