Learn More
Andwin Scientific 3 CARDBOARD FREEZER BOX 12/PK
SDP
Catalog No. 50727573
Encompass_Preferred
The 3" Cardboard Freezer Boxes and Dividers are suitable for Stainless Steel Freezer Racks. This product can hold cryogenic vials in liquid nitrogen or in mechanical freezers. The box footprint is 133 x 133 mm. This is sold in a pack of 12.
Catalog No. 50-727-573 Supplier Andwin Scientific Supplier No. 6161570
The 3" Cardboard Freezer Boxes and Dividers are suitable for Stainless Steel Freezer Racks. This product can hold cryogenic vials in liquid nitrogen or in mechanical freezers. The box footprint is 133 x 133 mm. This is sold in a pack of 12.