Learn More
Alkali Scientific Alkali Scientific™ 3" Cardboard Freezer Boxes with Drain Holes - Water and Ice Resistant Coating, No Dividers, 50/Case
SDP
Catalog No. 503029437
Encompass_Preferred
• Choose optional slot/cell dividers• Dividers also sold separately, item CD-• Temperature Range: -196 to +121• Drain holes in bottom corners• Writing area on the lid• Keeps tubes in an upright position for easy sorting Alkali Scientific™ 3" cardboard freezer box are ideal for any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cardboard Freezer Box - 5 1/4 x 5 1/4 x 3″ ( 133 x 133 x 75mm)
Catalog No. 50-302-9437 Supplier Alkali Scientific Supplier No. FB3CC50
• Choose optional slot/cell dividers• Dividers also sold separately, item CD-• Temperature Range: -196 to +121• Drain holes in bottom corners• Writing area on the lid• Keeps tubes in an upright position for easy sorting Alkali Scientific™ 3" cardboard freezer box are ideal for any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cardboard Freezer Box - 5 1/4 x 5 1/4 x 3″ ( 133 x 133 x 75mm)