Learn More
AdipoGen 3-CCA-Cl
SDP
Supplier:  AdipoGen CDXC0006M050
Encompass_Preferred
Chemical. CAS 196091-78-8. Formula C37H50O4. MW 558.79. Synthetic. Free coumarin-3-carboxylic acid shows a stronger fluorescence Em. max 419 nm Exc. 366 nm in ethanol.
Catalog No. 50-256-1564