Learn More
Cole-Parmer Traceable Triple-Display Digital Timer with Calibration
Catalog No. NC0741251 Shop All Cole-Parmer Products
Encompass_Preferred
Timer features three timer channels that can be used in any combination of up/down timing. Start all three channels at once or individually; distinct audible alarm sounds and "TIMES UP" flashes on the display when set time is reached. Memory function returns the display to the programmed time. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC0741251 Supplier Cole-Parmer Supplier No. 9446123
Timer features three timer channels that can be used in any combination of up/down timing. Start all three channels at once or individually; distinct audible alarm sounds and "TIMES UP" flashes on the display when set time is reached. Memory function returns the display to the programmed time. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.