Learn More
Checkers 3-Channel Rubber Duct Office Cable Protector
Catalog No. 19088437 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
19-088-437 Black
1 options
Catalog No. 19-088-437 Supplier Checkers Supplier No. RFD510
Only null left
Add to Cart
Add to Cart
Rubber duct systems protect and hide valuable cords and cables located in walkways or work areas. These non-metallic, flexible, industrial-grade rubber protectors are extremely durable, easy to install and organize portable cords and cables while reducing trip hazards.
  • Three-channel Protector for Multiple Cable Applications
  • Can Be Slit If Necessary (Supplied Unslit)
  • Reduces Tripping Hazards
  • Protects And Organizes Portable Cords And Cables
  • Non-Metallic, Industrial-Grade Rubber Construction
  • Multiple Sizes For Various Cable Diameters
  • Grooved Underside For Better Traction
  • Optional Connector Kit Available For Most Sizes
Specifications
Material Rubber
Product Type Cable Protector
Color Black
Length (English) 10 ft.
Length (Metric) 3 m