Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-1,2-benzisothiazole 98.0+%, TCI America™
SDP

Catalog No. C199125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C199125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C199125G Supplier TCI America Supplier No. C199125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7716-66-7
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
InChI Key BCPVKLRBQLRWDQ-UHFFFAOYSA-N
Synonym 3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole
PubChem CID 598190
IUPAC Name 3-chloro-1,2-benzothiazole
SMILES C1=CC=C2C(=C1)C(=NS2)Cl

Specifications

Melting Point 41°C
Color White-Yellow
Quantity 25 g
Formula Weight 169.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-1,2-benzisothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.