Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-3-Chloro-1,2-propanediol

Catalog No. 502898795
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-8795 500 mL
NC3365141 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-289-8795 Supplier Sigma Aldrich Supplier No. 107271500ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-chloro-1,2-propanediol undergoes enantioselective transformation to (r)-3-chloro-1,2-propanediol by the cell extract of corynebacterium sp
  • Strain n-1074
  • It is a contaminant found in food in free (diol) form as well as in an esterified (with fatty acids) form.(±)-3-chloro-1,2-propanediol may be used in manufacture of dye intermediates.3-chloro-1,2-propanediol is a potent rodent chemosterilant
TRUSTED_SUSTAINABILITY

Specifications

CAS 96-24-2
Density 1.322 g/mL (at 25°C (literature))
Boiling Point 213°C (lit.)
Molecular Formula C3H7ClO2
Refractive Index n20/D 1.480 (literature)
Linear Formula ClCH2CH(OH)CH2OH
MDL Number MFCD00004712
Quantity 500 mL
Synonym alpha-Chlorohydrin; alpha-Glycerol chlorohydrin; alpha-Monochlorohydrin; 3-MCPD
Molecular Weight (g/mol) 110.54
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.