Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-3-Chloro-1,2-propanediol, MilliporeSigma™ Supelco™

Catalog No. 111018731 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8731 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8731 Supplier MilliporeSigma Supelco Supplier No. 32406100MG
Add to Cart
Add to Cart

(+/-)-3-Chloro-1,2-propanediol is a potent human carcinogen. It is formed as a product of acid hydrolysis of vegetable proteins during the process of cooking.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 96-24-2
Vapor Pressure 0.04 mmHg (25°C)
Density 1.322 g/mL (at 25°C)
Flash Point 113°C - Closed Cup
Physical Form Neat
Linear Formula ClCH2 CH(OH)CH2 OH
MDL Number MFCD00004712
Boiling Point 213°C
Synonym alpha-Chlorohydrin; alpha-Glycerol chlorohydrin; alpha-Monochlorohydrin; 3-MCPD
Type Analytical Standard
Quantity 100 mg
Formula Weight 110.54
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.