Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,2-dimethyl-1-propanol 98.0+%, TCI America™
SDP

Catalog No. C153825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C153825G 25 g
C1538500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C153825G Supplier TCI America Supplier No. C153825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13401-56-4
Molecular Formula C5H11ClO
Molecular Weight (g/mol) 122.592
MDL Number MFCD00004684
InChI Key CAZPRAORHCOIHC-UHFFFAOYSA-N
PubChem CID 83407
IUPAC Name 3-chloro-2,2-dimethylpropan-1-ol
SMILES CC(C)(CO)CCl

Specifications

Color White
Quantity 25 g
Formula Weight 122.59
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Chloro-2,2-dimethyl-1-propanol
EINECSNumber (2)-2002
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.