Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific™

Catalog No. AAL2016603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2016603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2016603 Supplier Thermo Scientific Chemicals Supplier No. L2016603

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292621-58-0
Molecular Formula C7Cl2F4O
Molecular Weight (g/mol) 246.97
MDL Number MFCD03001175
InChI Key GNXAQPALNSLOBK-UHFFFAOYSA-N
Synonym acmc-1cptj, 3-chloro-2,4,5,6-tetrafluorobenzoylchloride, benzoyl chloride,3-chloro-2,4,5,6-tetrafluoro, 3-chloranyl-2,4,5,6-tetrakis fluoranyl benzoyl chloride
PubChem CID 2783249
IUPAC Name 3-chloro-2,4,5,6-tetrafluorobenzoyl chloride
SMILES FC1=C(F)C(Cl)=C(F)C(C(Cl)=O)=C1F

Specifications

Refractive Index 1.4898
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 246.99
Percent Purity 98%
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.