Learn More
Medchemexpress LLC 3-chloro-2,4-difluorobenzoic acid | 154257-75-7 | MFCD01631387 | 99.9% | 192.55 g·mol^-1 | C7H3ClF2O2 | 50 G

Supplier: Medchemexpress LLC HYW01508250G
3-Chloro-2,4-difluorobenzoic acid is an organic biochemical reagent used as an intermediate in medicinal chemistry and life-science research. Supplied as a high-purity powder, it is appropriate for synthesis, analytical characterization, and method development. The product includes recommended storage conditions to maximize stability and shelf life.
- High purity (99.94%) for reliable synthetic and analytical results.
- Molecular formula C7H3ClF2O2 and molecular weight 192.55 g·mol^-1 for accurate stoichiometry.
- Available in multiple pack sizes, including 50 g, to suit bench-scale work.
- Powder form; store at -20°C for long-term stability or 4°C for shorter-term storage.
- Suitable as an intermediate in medicinal chemistry and method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.