Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,5-dimethylpyrazine 95.0+%, TCI America™
SDP

Catalog No. C20405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20405G Supplier TCI America Supplier No. C20405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95-89-6
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00006143
InChI Key NNBALVIZMGWZHS-UHFFFAOYSA-N
PubChem CID 66771
IUPAC Name 3-chloro-2,5-dimethylpyrazine
SMILES CC1=CN=C(C)C(Cl)=N1

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 142.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2,5-dimethylpyrazine
RTECSNumber UQ2462000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.