Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C29461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29461G 1 g
C2946-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29461G Supplier TCI America Supplier No. C29461G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 3-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 352535-82-1
Melting Point 247°C
Color White
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD05664224
Synonym 3-chloro-2-fluorophenyl boronic acid, 3-chloro-2-fluorobenzeneboronic acid, boronic acid, 3-chloro-2-fluorophenyl, boronic acid,b-3-chloro-2-fluorophenyl, acmc-209ifa, 3-chloro-2-fluorophenyl boranediol, 3-chloro-2-fluorophenyl boronicacid, 2-fluoro-3-chlorophenylboronic acid, 3-chloro-2-fluorophenylboronic acid
InChI Key SUYRGLRWMPEARP-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC(Cl)=C1F
IUPAC Name (3-chloro-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.36
PubChem CID 16217158
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.