Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Chloro-2-Hydroxypropyl)Trimethylammonium Chloride Solution

Catalog No. 3482871L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-828-71L 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 34-828-71L Supplier Sigma Aldrich Supplier No. 3482871L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution (CHTAC) can be used:As a cation-generating agent for cellulose cationization by exhaustion method.To resolve 2,2'sa-dihydroxy-1,1'sa-binaphthyl enantiomers.To synthesize cationic glycogen (Cat Gly).As a quaternizing agent for quaternization of N-aryl chitosan derivatives.

Specifications

CAS 64536-78-3
Molecular Formula C8H12Cl2Ru ]n
Linear Formula [Ru(COD)Cl2]n
Refractive Index n20/D 1.4541
Quantity 1 g
Synonym (1,5-Cyclooctadiene)ruthenium(II) chloride; Ruthenium(II) chloride 1,5-cyclooctadiene complex
Molecular Weight (g/mol) 280.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.