Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-hydroxypyridine, 97%

Catalog No. AAL2002703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2002703 1 g
AAL2002706 5 g
AAL2002714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2002703 Supplier Thermo Scientific Chemicals Supplier No. L2002703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13466-35-8 | C5H4ClNO | 129.54 g/mol

3-Chloro-2-hydroxypyridine is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-2-hydroxypyridine is used as an organic intermediate.

Solubility
Soluble in water (25 deg C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13466-35-8
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD01646109
InChI Key XTYNIPUFKBBALX-UHFFFAOYSA-N
Synonym 3-chloro-2-hydroxypyridine, 3-chloropyridin-2-ol, 3-chloro-2-pyridinol, 2-hydroxy-3-chloropyridine, 3-chloropyridin-2 1h-one, 3-chloro-2 1h-pyridinone, chloropyridone, 2 1h-pyridinone, 3-chloro, chloropyridinol, pubchem6538
PubChem CID 582210
IUPAC Name 3-chloro-1H-pyridin-2-one
SMILES ClC1=CC=CNC1=O

Specifications

Melting Point 180°C to 182°C
Quantity 1 g
Beilstein 109845
Solubility Information Soluble in water (25 deg C)
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.